Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Ms-3 (Mouse), 4.60 mm, 100 mm
CA76204-856
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
: For research use only.
- Ø int.:4.60 mm
- Désignation:Multiple Affinity Removal Column - Ms-3 (Mouse)
- Longueur:100 mm



