Pre-Printed Laboratory Warning Labels
Product Details & Documents
Various pre-printed laboratory warning labels designed to advise staff and visitors of potential hazards and dangers. This includes biohazard and radioactive warning labels, as well as standardized THC warning labels for labeling a wide range of cannabis products.
- Temperature range: −80 to +100 °C (−112 to +212 °F)
- Withstands ultra-low temperature conditions (−80 °C/−112 °F)
- Special construction protects against abrasion, chemical exposure, and tearing
- Durable and high-quality materials for long-lasting use
Pre-printed general warning labels for identifying equipment, containers, and material that may pose a safety risk in the laboratory environment. With a wide temperature resistance range, these labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, etc.), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts.
Spécifications
- Color:Black on yellow
- Désignation:Warning labels - Radioactive
- Dimensions:12.7 mm (0.5″)
- Matière:Plastic
- Packaging:1000 labels/roll (1 label across)
- Core Size:3"
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



