Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cryogenic Lamination Tapes | Avantor

Cryogenic Lamination Tapes

Cryogenic lamination tape, Transparent, 3", 50.8×60000 mm (2×2400")
Catalog # 76276-002
Fournisseur #:  807656005881
Cryogenic Lamination Tapes
Catalog # 76276-002
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes

Cryogenic lamination tape, Transparent, 3", 50.8×60000 mm (2×2400")

Price
$108.00
Log in pour la disponibilité
Roll of 1
Product Details & Documents

Thin cryo lamination tape provides added protection against abrasion, harsh chemicals, and extreme temperatures. Designed to withstand long-term storage in liquid nitrogen and ultra-low temperature freezers, multiple freeze-thaw cycles, and transport on dry ice.

  • Temperature range: −196 to +93 °C (−321 to +199 °F)
  • Clear lamination tape protects material from extreme cryogenic conditions, including liquid phase and vapor phase liquid nitrogen
  • Resists harsh laboratory environments, including multiple freeze-thaw cycles
  • Offers protection against chemicals and solvents
  • Resistant to gamma irradiation (tested up to 50 KGy)

This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as 196 °C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.

Spécifications
  • Color:
    Transparent
  • Désignation:
    Cryogenic lamination tape
  • Dimensions:
    50.8×60000 mm (2×2400")
  • Core Size:
    3"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.