Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Cryogenic Color Dots Writable Labels

Cryogenic color dots writable label, Red, 19 mm (0.75"), 420 labels/pack (21 labels/sheet)
  76275-678
 :  807656025254
Cryogenic Color Dots Writable Labels
  76275-678
Cryogenic Color Dots Writable Labels
Cryogenic Color Dots Writable Labels

Cryogenic color dots writable label, Red, 19 mm (0.75"), 420 labels/pack (21 labels/sheet)

 
$38.08
   
Pack of 1
 

These small diameter, round cryo labels for microtube tops are made with a strong adhesive. Ideal for −80 °C freezers, dry ice, and liquid nitrogen storage.

  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Available in a wide range of colors, for color-coding and easy identification
  • Easy to write on with permanent markers
  • Small round labels are ideal for 0.5 to 1.5 ml Eppendorf tube caps and vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Resistant to gamma irradiation (tested to up to 50 kGy)

Specially designed to comfortably fit 0.5, 0.6, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors, as well as packs of 15 assorted colors for color-coding and rapid identification.

 
  • Color:
    Red
  • Désignation:
    Cryogenic color dots writable label
  • Dimensions:
    19 mm (0.75")
  • Packaging:
    420 labels/pack (21 labels/sheet)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.