Self-Laminating Calibration Labels
Product Details & Documents
Self-laminating calibration labels for secure and safe identification of regularly inspected equipment and instruments. These labels can be inscribed with all relevant information and then laminated with the supplied clear laminate to provide physical protection against various abuses.
- Temperature range: −80 to +93 °C (−112 to +200 °F)
- Can withstand extreme temperatures, including cryogenic and high heat conditions
- Resists abrasion and exposure to harsh chemicals and alcohols
- Ideal for use with equipment that will undergo preventative maintenance, calibration, or re-certification
- Once laminated the information is protected and cannot be altered
Self-laminating, writable, calibration labels designed for equipment that is calibrated or inspected regularly. The labels can be inscribed using permanent ink markers. The clear laminate is applied over the label, protecting the written information. These labels can resist abrasion and exposure to harsh chemicals, such as ethanol, acetone, isopropanol, thinners, detergents, and surface sanitizing wipes.
The self-laminating calibration labels are ideal for labeling regularly inspected medical equipment, equipment that has undergone preventative maintenance, that require calibration prior to use, or that are out of order and must not be used. Reagent calibration labels are also available for time-sensitive solutions prepared in the lab. Calibration label customization is also available to include the company's logos and information.
Spécifications
- Color:Black on white
- Désignation:Self-laminating calibration labels
- Dimensions:25.4×54 mm (1×2.125")
- Matière:Plastic
- Packaging:1000 labels/roll (1 label across)
- Core Size:3"
- Legend:—
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



