dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific | Avantor
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipette, with US Plug, 100 - 1000 µl, 5 µl
Catalog # 76271-364
Fournisseur #:  7016201004
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
Catalog # 76271-364
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipette, with US Plug, 100 - 1000 µl, 5 µl
Price
$1,007.13
Log in pour la disponibilité
Each
Product Details & Documents
High accuracy, high performance stepper motor ensures accuracy and repeatability, eliminating manual pipetting errors.
  • Two buttons handle all operational settings
  • Lightweight, ergonomic design
  • Small body dimensions enable easy handling and fatigue-free pipetting
  • Adjustable speeds for aspiration and dispensing
  • Li-ion battery and dual charging modes enable longer operation time
  • Self calibration via USB to PC; third party calibration is not required
A motor driven digital control pipette with multifunctional capabilities of pipetting, mixing, stepper, and dilution.
Electrical: 100 to 220 V, 50/60 Hz
Certifications: CE marked. cTUVus.
Delivery information: Each package includes the electronic pipette with battery, stand, charger, USB cable, calibration card, and user manual.
Spécifications
  • Volume:
    100 - 1000 µl
  • Désignation:
    dPette+ Electronic Pipette, with US Plug
  • Increments:
    5 µl