dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipette, with US Plug, 0.5 - 10 µl, 0.01 µl
Catalog # 76271-358
Supplier: DLAB SCIENTIFIC INC.
Fournisseur #: 7016201001
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
Catalog # 76271-358
Supplier: DLAB SCIENTIFIC INC.
Jump to:
Product Details & Documents
High accuracy, high performance stepper motor ensures accuracy and repeatability, eliminating manual pipetting errors.
- Two buttons handle all operational settings
- Lightweight, ergonomic design
- Small body dimensions enable easy handling and fatigue-free pipetting
- Adjustable speeds for aspiration and dispensing
- Li-ion battery and dual charging modes enable longer operation time
- Self calibration via USB to PC; third party calibration is not required
A motor driven digital control pipette with multifunctional capabilities of pipetting, mixing, stepper, and dilution.
Electrical: 100 to 220 V, 50/60 Hz
Certifications: CE marked. cTUVus.
Delivery information: Each package includes the electronic pipette with battery, stand, charger, USB cable, calibration card, and user manual.
Spécifications
- Volume:0.5 - 10 µl
- Désignation:dPette+ Electronic Pipette, with US Plug
- Increments:0.01 µl