Catheterization Trainers
Product Details & Documents
Fits most laerdal manikins.
Cathi is a simulated female catheterization trainer. The tissue is supple and pliable for easy spreading using the thumb and index finger. The labia minora realistically closes over the vaginal and urethral openings. Use to practice locating the urethral meatus and urinary catheter placement. Willi is a simulated male catheterization trainer. Willi fits into most Laerdal manikins or into the Remedy stand. The feel of the material is realistic and delicate like actual tissue. Willi has a removable foreskin and can simulate an enlarged prostate. Each is shipped with a practice catheter.
Successful catheterization on either will be confirmed with urine flashback. The bladder module is used with an IV bag (not Included) that can be hung or inserted into a pressure infuser bag (sold separately) to provide flashback.
Ordering information: Willi and Cathi fit into most Laerdal manikins or into the Remedy Stand (sold separately). Does Not include Torso.
Specifications
- Désignation:Willi Model