FectoCHO® Expression System
FectoCHO® Expression System
FectoCHO™ CD Expression Medium, 6×1 L (Expression Medium), —
Catalog # CA76234-742
Supplier #:  101000003
FectoCHO® Expression System
Catalog # CA76234-742
CAS Number:  
FectoCHO® Expression System
FectoCHO® Expression System
FectoCHO™ CD Expression Medium, 6×1 L (Expression Medium), —
CAS Number:  
Price
$1,723.56
Log in pour la disponibilité
Each (6l)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
FectoCHO® Expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD Expression medium and a powerful transfection reagent, FectoPRO®.
  • Outstanding protein production yields in various CHO cells
  • Fast and effortless CHO cell adaptation
  • Chemically defined kit components
  • Excellent lot to lot reproducibility
  • Cost-effective solution
FectoCHO® CD Expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with the competitor's systems.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Ordering information: FectoPRO® transfection reagent, FectoPRO® Booster and FectoCHO® CD Expression Medium can be purchased separately.
Spécifications
Spécifications
  • Désignation:
    FectoCHO™ CD Expression Medium
  • Taille:
    6×1 L (Expression Medium)
  • Buffer Volume:
  • Cat. No.:
    CA76234-742
  • Cell specific:
    CHO
  • Bioproduction:
    Yes