Multivapor™ Parallel Evaporation Systems, Buchi | Avantor
Multivapor™ Parallel Evaporation Systems, Buchi
Uses Rotavapor®, 12
Catalog # 76266-284
Multivapor™ Parallel Evaporation Systems, Buchi
Catalog # 76266-284
Supplier:  
Multivapor™ Parallel Evaporation Systems, Buchi
Multivapor™ Parallel Evaporation Systems, Buchi
Uses Rotavapor®, 12
Discontinued
Product Details & Documents
Evaporate up to 12 samples simultaneously by efficient vacuum vortex technology.
  • Fast evaporation of multiple samples
  • Easy operation
  • Integration with rotavapor
  • Full visibility of samples
Speed up your sample evaporation for multiple samples. With its ease of use, the Multivapor™ is tailored to maximize efficiency. A wide range of available adapters allow neat integration of your glassware into the daily routine.
Spécifications
  • Condenser Type:
    Uses Rotavapor®
  • No. of Positions:
    12