Multivapor™ Parallel Evaporation Systems, Buchi
Uses Rotavapor®, 12
Catalog # 76266-284
Jump to:
Discontinued
Product Details & Documents
Evaporate up to 12 samples simultaneously by efficient vacuum vortex technology.
- Fast evaporation of multiple samples
- Easy operation
- Integration with rotavapor
- Full visibility of samples
Speed up your sample evaporation for multiple samples. With its ease of use, the Multivapor™ is tailored to maximize efficiency. A wide range of available adapters allow neat integration of your glassware into the daily routine.
Spécifications
- Condenser Type:Uses Rotavapor®
- No. of Positions:12