Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 45/50, 24/40
  76264-370
 :  F43545L
Synthware Round-Bottom Flasks with Three Necks, Angled
  76264-370
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 45/50, 24/40
$588.26
Each
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacité:
    5000 ml
  • Center Neck [ST] Joint:
    45/50
  • Side Neck ST Joints:
    24/40