Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 34/45, 29/42
Catalog # 76264-368
Supplier:   Kemtech America
Supplier #:  F43385L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-368
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 34/45, 29/42
Price
$575.65
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    5000 ml
  • Center Neck [ST] Joint:
    34/45
  • Side Neck ST Joints:
    29/42