Synthware Round-Bottom Flasks with Four Necks
5000 ml, 45/50, 29/42
Catalog # 76264-128
Supplier: Kemtech America
Supplier #: F21585L
Jump to:
Product Details & Documents
With identical vertical side joints.
- 3 Straight side necks
- Ground glass joints
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Specifications
- Capacité:5000 ml
- Center Neck [ST] Joint:45/50
- Side Neck ST Joints:29/42