Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
5000 ml, 45/50, 29/42
Catalog # 76264-128
Supplier:   Kemtech America
Supplier #:  F21585L
Synthware Round-Bottom Flasks with Four Necks
Catalog # 76264-128
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
5000 ml, 45/50, 29/42
Price
869,75 $
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Specifications
  • Capacité:
    5000 ml
  • Center Neck [ST] Joint:
    45/50
  • Side Neck ST Joints:
    29/42