Synthware Round-Bottom Flasks with Four Necks | Avantor
Synthware Round-Bottom Flasks with Four Necks
5000 ml, 45/50, 24/40
Catalog # 76264-126
Supplier:   Kemtech America
Fournisseur #:  F21545L
Synthware Round-Bottom Flasks with Four Necks
Catalog # 76264-126
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
5000 ml, 45/50, 24/40
Price
$861.35
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Spécifications
  • Capacité:
    5000 ml
  • Center Neck [ST] Joint:
    45/50
  • Side Neck ST Joints:
    24/40