Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 29/42, 29/42
Catalog # 76264-098
Supplier:   Kemtech America
Supplier #:  F43295L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-098
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
5000 ml, 29/42, 29/42
Price
$561.37
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacité:
    5000 ml
  • Center Neck [ST] Joint:
    29/42
  • Side Neck ST Joints:
    29/42