Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 29/42, 29/42
Catalog # 76264-084
Supplier: Kemtech America
Fournisseur #: F43293L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-084
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:3000 ml
- Center Neck [ST] Joint:29/42
- Side Neck ST Joints:29/42