Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
3000 ml, 29/42, 29/42
Catalog # 76264-058
Supplier:   Kemtech America
Supplier #:  F41823L
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 76264-058
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
3000 ml, 29/42, 29/42
Price
$384.07
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
  • Angled side neck
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
  • Capacité:
    3000 ml
  • Center Neck [ST] Joint:
    29/42
  • Side Neck ST Joints:
    29/42