Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
3000 ml, 29/42, 29/42
Catalog # 76264-058
Supplier: Kemtech America
Supplier #: F41823L
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 76264-058
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
- Angled side neck
- Heavy wall borosilicate construction
- Ground glass joint
- Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Spécifications
- Capacité:3000 ml
- Center Neck [ST] Joint:29/42
- Side Neck ST Joints:29/42