Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Connectors, Digi-Key

Connectors, Digi-Key

Connector Header Surface Mount, 20 Positions 0.050" (127 mm)
Catalog # 76252-744
Supplier #:  FTSH-125-01-F-
Connectors, Digi-Key
Catalog # 76252-744
Connectors, Digi-Key

Connector Header Surface Mount, 20 Positions 0.050" (127 mm)

Price
55,06 $
Log in for availability
Each
Product Details & Documents
Specifications
  • Désignation:
    Connector Header Surface Mount, 20 Positions 0.050" (127 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.