Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Connectors, Digi-Key

Connectors, Digi-Key

Connector Header Vertical, 20 Positions 0.050" (127 mm)
Catalog # 76252-522
Supplier #:  104549-2
Connectors, Digi-Key
Catalog # 76252-522
Connectors, Digi-Key

Connector Header Vertical, 20 Positions 0.050" (127 mm)

Price
32,88 $
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    Connector Header Vertical, 20 Positions 0.050" (127 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.