TE Connectivity Crimping Tools, Mouser Electronics
Crimper Wire, Female
Catalog # 76251-508
Supplier: MOUSER ELECTRONICS TE
Fournisseur #: 217885-1
TE Connectivity Crimping Tools, Mouser Electronics
Catalog # 76251-508
Supplier: MOUSER ELECTRONICS TE
Jump to:
Product Details & Documents
These crimping tools are designed with key crimping tool characteristics to enhances the overall performance of the production termination process.
- Designed with high wear resistant materials
Terminals are designed to perform to specification only when the final crimp form is within a narrow range of dimensions.
Spécifications
- Désignation:Crimper Wire, Female