Bottoming Chamfer, Sid Tool
Bottoming Chamfer Three Flutes, Heli-Coil, High Speed Steel, 2⅜", Bright, 0.2200", ¹⁵⁄₁₆"
76251-130
: 64832033
Bottoming chamfers are used for threading blind holes to the bottom. They have 1 to 2 threads. Bright provides a smooth, polished finish on the tool. It increases chip flow in softer materials such as aluminum, wood and plastic.
High Speed Steel (HSS) provides good wear-resistance and can be used in general purpose applications for both ferrous and non-ferrous materials.
- Désignation:Bottoming Chamfer Three Flutes, Heli-Coil
- Longueur:2⅜"
- Matière:High Speed Steel
- Finish:Bright
- Shank Diameter:0.2200"
- Thread Length:¹⁵⁄₁₆"