Retainer System and Cap Assembly, EFD | Avantor
Retainer System and Cap Assembly, EFD
Retainer System, with No Regulator, 6 oz
Catalog # 76248-690
Fournisseur #:  7012433
Retainer System and Cap Assembly, EFD
Catalog # 76248-690
Retainer System and Cap Assembly, EFD
Retainer System, with No Regulator, 6 oz
Price
$1,150.98
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Retainer system includes cartridges, pistons, outlet caps, end caps and retainers. For use with most assembly fluids and pastes. Sold individually.
Spécifications
  • Désignation:
    Retainer System, with No Regulator
  • Taille:
    6 oz