Envelope Gaskets, Garlock Hygienic Technologies
Envelope gasket, PTFE, 2.0"
Catalog # 76246-012
Supplier: Rubber Fab
Fournisseur #: A40MPGR-200-V
Jump to:
Product Details & Documents
A PTFE envelope gasket has the chemical resistance of PTFE and the elastomeric characteristics of FKM Fluoroelastomer.
Certifications: Meets FDA and USP Class VI standards.
Spécifications
- Désignation:Envelope gasket
- Matière:PTFE
- Taille:2.0"