Envelope Gaskets, Garlock Hygienic Technologies | Avantor
Envelope Gaskets, Garlock Hygienic Technologies
Envelope gasket, PTFE, 2.0"
Catalog # 76246-012
Supplier:   Rubber Fab
Fournisseur #:  A40MPGR-200-V
Envelope Gaskets, Garlock Hygienic Technologies
Catalog # 76246-012
Supplier:  Rubber Fab
Envelope Gaskets, Garlock Hygienic Technologies
Envelope gasket, PTFE, 2.0"
Price
$13.33
Log in pour la disponibilité
Each
Product Details & Documents
A PTFE envelope gasket has the chemical resistance of PTFE and the elastomeric characteristics of FKM Fluoroelastomer.
Certifications: Meets FDA and USP Class VI standards.
Spécifications
  • Désignation:
    Envelope gasket
  • Matière:
    PTFE
  • Taille:
    2.0"