Workspace Organization Tools, Production Basics
Overhead Arm, Pair, —, 9 lbs.
  76245-008
 :  PB-8538
Workspace Organization Tools, Production Basics
  76245-008
Workspace Organization Tools, Production Basics
Workspace Organization Tools, Production Basics
Overhead Arm, Pair, —, 9 lbs.
$493.20
Each
The cable troughs are designed to store cables for easier access and to improve wire management on the workstations.
Mounting arms allow the overhead light and Tool track trolley to be attached to your work bench. Compatible with the upright open frame and uprights.
Cable troughs are mounted to the upright frame and are available in four standard sizes.
  • Désignation:
    Overhead Arm, Pair
  • Weight:
    9 lbs.
  • Width: