Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Single Flute Countersink, Cutting Tool Supply

Single Flute Countersink, 90°, 1", 2¾", ½"
Catalog # 76244-654
 :  C46132
Single Flute Countersink, Cutting Tool Supply
Catalog # 76244-654
Single Flute Countersink, Cutting Tool Supply

Single Flute Countersink, 90°, 1", 2¾", ½"

Price
$419.66
Log in pour la disponibilité
Each
Product Details & Documents

Made of black oxide with reduced shank.

 : ASME B94.11M.

Spécifications
  • Désignation:
    Single Flute Countersink, 90°
  • Longueur:
    2¾"
  • Shank Diameter:
    ½"
  • Diameter:
    1"