Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Single Flute Countersink, Cutting Tool Supply

Single Flute Countersink, Cutting Tool Supply

Single Flute Countersink, 100°, 1", 2¾", ½"
  76244-644
Supplier #:  C46133
Single Flute Countersink, Cutting Tool Supply
  76244-644
Single Flute Countersink, Cutting Tool Supply

Single Flute Countersink, 100°, 1", 2¾", ½"

 
$426.73
   
Each
 

Made of black oxide with reduced shank.

Certifications: ASME B94.11M.

 
  • Désignation:
    Single Flute Countersink, 100°
  • Longueur:
    2¾"
  • Shank Diameter:
    ½"
  • Diameter:
    1"