Dispensing Tips, EFD INC.
25, Standard, Red
Supplier: EFD INC.- CUSTOMER SERVICE TE
Supplier #: 7018391
Jump to:
Product Details & Documents
Smooth flow for application of medium- to high-viscosity fluids especially thick or particle-filled materials like epoxies, RTVs and braze pastes.
- Free of flash, burrs and contaminants
- Package labels include lot numbers for process control and traceability
- Consistent from style to style and lot to lot
- 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel
- Engineered hub flats for easy twist on, twist off
Molded of polyethylene with UV-light block additive.
Packaging: Packaged 50 tips per box.
Spécifications
- Color:Red
- Point Style:Standard
- Jauge:25