Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

1X TBS Buffer with 1% Casein

1X TBS Buffer with 1% Casein, 1 L
Catalog # 76236-548
 :  MB-082-1000
1X TBS Buffer with 1% Casein
Catalog # 76236-548
CAS Number:  
1X TBS Buffer with 1% Casein

1X TBS Buffer with 1% Casein, 1 L

CAS Number:  
Price
221,78 $
Log in pour la disponibilité
Each (1l)
 
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Tris buffered saline with casein is suitable for multiple applications including biological diluent buffer for antibodies or other biologics.

  • Ready-to-Use
  • Diluent or wash buffer for assays
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria

It also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.

Application Note: This product is a ready-to-use 1X solution and no further dilution is required.

Spécifications

Spécifications

  • Désignation:
    1X TBS Buffer with 1% Casein
  • Taille:
    1 L

Spécifications