Large Scale Evaporator Flasks, Buchi®
20 L, R-153/R-220, Evaporating flask, 150 mm
Catalog # CA89034-512
Supplier: BUCHI CORPORATION CA
: 027468
Jump to:
Product Details & Documents
Standard glass flask for use with various Rotavapor™ Evaporator models.
Spécifications
- Capacité:20 L
- Désignation:Evaporating flask
- Flange O.D.:150 mm
- Fits Models:R-153/R-220