VWR® Cap Mat
96-Well Round Well Cap Mat, EVA, Non sterile
Catalog # 76221-326
Supplier: VWR International
Jump to:
Product Details & Documents
Re-usable cap mats are available for friction sealing of plates.
Cap mats are an economic alternative to thermal or adhesive seals as they can be sterilized and reused.
Spécifications
- Sterility:Non sterile
- Désignation:96-Well Round Well Cap Mat
- Matière:EVA