Sling Psychrometer
Sling Psychrometer, Pack of 15
Catalog # 470304-042
Supplier: United Scientific Supplies
Fournisseur #: SPCRKIT-PK/15
Jump to:
Product Details & Documents
Use this simple apparatus to measure relative humidity.
- Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Spécifications
- Désignation:Sling Psychrometer, Pack of 15