Sling Psychrometer | Avantor
Sling Psychrometer
Sling Psychrometer, Pack of 15
Catalog # 470304-042
Fournisseur #:  SPCRKIT-PK/15
Sling Psychrometer
Catalog # 470304-042
Sling Psychrometer
Sling Psychrometer
Sling Psychrometer, Pack of 15
Price
165,90 $
Log in pour la disponibilité
Pack of 15
Product Details & Documents
Use this simple apparatus to measure relative humidity.
  • Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Spécifications
  • Désignation:
    Sling Psychrometer, Pack of 15