Sling Psychrometer | Avantor
Sling Psychrometer
Sling Psychrometer
Catalog # 470304-040
Fournisseur #:  SPCRKIT
Sling Psychrometer
Catalog # 470304-040
Sling Psychrometer
Sling Psychrometer
Sling Psychrometer
Price
$23.85
Log in pour la disponibilité
Each
Product Details & Documents
Use this simple apparatus to measure relative humidity.
  • Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Spécifications
  • Désignation:
    Sling Psychrometer