Sling Psychrometer
Sling Psychrometer
Catalog # 470304-040
Supplier: United Scientific Supplies
Fournisseur #: SPCRKIT
Jump to:
Product Details & Documents
Use this simple apparatus to measure relative humidity.
- Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Spécifications
- Désignation:Sling Psychrometer
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.