Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Secure specimens during processing with the Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
: CE-IVD.
: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
- Color:Orange