Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Aqua
Catalog # CA76214-148
Supplier:   Simport Scientific
Supplier #:  M510-12T
Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Catalog # CA76214-148
Supplier:  Simport Scientific
Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Aqua
Price
$1,404.18
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Slimsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvent
 : CE-IVD.
 : Each case contains 50 Quickload™ stacks of 40 cassettes.
Specifications
  • Color:
    Aqua