Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Lilac
Catalog # CA76214-124
Supplier:   Simport Scientific
 :  M507-10T
Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Catalog # CA76214-124
Supplier:  Simport Scientific
Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Micromesh™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Lilac
Price
$1,404.18
Log in pour la disponibilité
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Micromesh™ Biopsy Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 45° anterior writing area
  • 1676 square openings (0.36 mm)
  • No biopsy pads needed
  • One large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
 : CE-IVD.
 : Each case contains 50 Quickload™ stacks of 40 cassettes.
Spécifications
  • Color:
    Lilac