Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Product Details & Documents
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
- In Quickload™ stacks
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevents early separation of base and lid
- Snap latch and hinge-lock design; cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.
Spécifications
- Color:Gray