Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Orange
Catalog # CA76214-078
Supplier:   Simport Scientific
Supplier #:  M492-11T
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # CA76214-078
Supplier:  Simport Scientific
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Orange
Price
$986.23
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Histosette® II Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Open and close as often as necessary; cassettes will always relock securely
  • Back-mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Specifications
  • Color:
    Orange