Histosette® II Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Secure specimens during processing with the Histosette® II Tissue Processing/Embedding Cassette.
- In Quickload™ stacks
- 45° anterior writing area
- Open and close as often as necessary; cassettes will always relock securely
- Back-mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
- Color:Pink