BioClean™ Microflow Face Veil with Headloop, Nitritex
White, Non sterile, 330 ±5 mm, 380 ±5 mm
CA75873-948
: BFV05
BioClean™ Microflow Cleanroom Face Veil is low-linting with ultrasonically sealed edges reducing the risk of contamination into the controlled environment.
- Veil made from low linting thermabond non-woven material
- White hydroentangled polyester binding and tubular polyester head loop
- Processed in a cleanroom environment, contains no latex
Features a fully enclosed malleable aluminum nose-band with plastic coated and head-loops for a good fit. Suitable for use in class 10, ISO Class 4, M2.5 cleanrooms.
: Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
: 100 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; 10 outer bags per lined carton (1000/carton).
- Sterility:Non sterile
- Largeur:330 ±5 mm
- Color:White
- Longueur:380 ±5 mm