Rig Lizard® 2030 Gloves, HexArmor
  76207-902
Rig Lizard® 2030 Gloves, HexArmor
  76207-902
 :  
Rig Lizard® 2030 Gloves, HexArmor
Rig Lizard® 2030 Gloves, HexArmor
The Rig Lizard® 2030 delivers maximum grip and dexterity without sacrificing protection. It features a hi-vis silicone palm for superior grip and heat resistance, and the patented high-flex IR-X® Impact Exoskeleton™ protects the entire back- of-hand from smash and pinch injuries.
  • SuperFabric® brand material palm provides ANSI/ISEA cut level A6 resistance for some of the highest cut protection available in the industry (interior layer)
  • Glove was tested to the EN407 standard and received a level 3
  • Durable synthetic leather palm with heat-resistant silicone pattern provides superior grip and dexterity
  • Back-of-hand IR-X® Impact Exoskeleton™ with high-flex design keeps hands safe from smash and impact hazards
  • SlipFit® cuff with Velcro® closure for a secure and comfortable fit
  • Hi-vis color scheme increases visual awareness
  • Launderable
The Rig Lizard® 2030 with A6 cut resistance is a leader in the oil, gas, and mining industries. The durable TP-X® grip technology provides the ultimate grip in wet/dry applications while the patented full back-of-hand IR-X® Impact Exoskeleton™ keeps hands safe from smash and pinch hazards.
  • Cat. No.:
    76207-902
  • EN norm:
    En388 4532