Rig Lizard® 2030 Gloves, HexArmor
76207-902
The Rig Lizard® 2030 delivers maximum grip and dexterity without sacrificing protection. It features a hi-vis silicone palm for superior grip and heat resistance, and the patented high-flex IR-X® Impact Exoskeleton™ protects the entire back- of-hand from smash and pinch injuries.
- SuperFabric® brand material palm provides ANSI/ISEA cut level A6 resistance for some of the highest cut protection available in the industry (interior layer)
- Glove was tested to the EN407 standard and received a level 3
- Durable synthetic leather palm with heat-resistant silicone pattern provides superior grip and dexterity
- Back-of-hand IR-X® Impact Exoskeleton™ with high-flex design keeps hands safe from smash and impact hazards
- SlipFit® cuff with Velcro® closure for a secure and comfortable fit
- Hi-vis color scheme increases visual awareness
- Launderable
The Rig Lizard® 2030 with A6 cut resistance is a leader in the oil, gas, and mining industries. The durable TP-X® grip technology provides the ultimate grip in wet/dry applications while the patented full back-of-hand IR-X® Impact Exoskeleton™ keeps hands safe from smash and pinch hazards.
- Cat. No.:76207-902
- EN norm:En388 4532