MX200 Protective Eyewear, HexArmor | Avantor
MX200 Protective Eyewear, HexArmor
Protective eyewear MX200, Anti-fog, scratch-resistant/hardcoat (TruShield® 2F), Clear
Catalog # 76207-752
Supplier:   HexArmor
Fournisseur #:  11-10002-03
MX200 Protective Eyewear, HexArmor
Catalog # 76207-752
Supplier:  HexArmor
MX200 Protective Eyewear, HexArmor
MX200 Protective Eyewear, HexArmor
Protective eyewear MX200, Anti-fog, scratch-resistant/hardcoat (TruShield® 2F), Clear
Price
$31.02
Log in pour la disponibilité
Each
Product Details & Documents
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.Dual-injection soft-grip side arms for comfort and non-slip fit.
  • Rimless lens design with wide field of vision
  • Integrated channel for eye ventilation
  • Beveled lens edge for additional upper eye protection
  • Contoured nose bridge for comfort and fit
  • LTX™ Lens Technology for premium optical clarity
  • 100% UV protection
  • Metal-free hinge design
The MX200 with permanent anti-fog coatings, features a wraparound lens design that delivers maximum protection, the MX200 has a durable frame with a soft, low-pressure grip thanks to the unique dual injection technology.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.Attachable foam guard available.
Certifications: Meets Standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.
Spécifications
  • Désignation:
    Protective eyewear MX200
  • Lens Coating:
    Anti-fog, scratch-resistant/hardcoat (TruShield® 2F)
  • Lens Tint:
    Clear