Multiple Affinity Removal Column Human 14, Agilent Technologies
Multiple Affinity Removal Column, Human-14, 4.60 mm, 50 mm
Catalog # CA76193-736
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
: 5188-6557
Multiple Affinity Removal Column Human 14, Agilent Technologies
Catalog # CA76193-736
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Spécifications
- Ø int.:4.60 mm
- Désignation:Multiple Affinity Removal Column, Human-14
- Longueur:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



