SureVector, Vectors, Agilent Technologies
SureVector E. coli N-terminal tag kit, 5 reactions, SureVector E. coli N-terminal tag Kit E. coli starter kit containing 3 separate N-terminal tags and a T7 promoter
Catalog # CA76193-682
SureVector, Vectors, Agilent Technologies
Catalog # CA76193-682
Supplier:  
CAS Number:  
SureVector, Vectors, Agilent Technologies
SureVector, Vectors, Agilent Technologies
SureVector E. coli N-terminal tag kit, 5 reactions, SureVector E. coli N-terminal tag Kit E. coli starter kit containing 3 separate N-terminal tags and a T7 promoter
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
  • Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
  • Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
  • More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
  • Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the Agilent set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
 : Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
 : For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
  • Désignation:
    SureVector E. coli N-terminal tag kit
  • Taille:
    5 reactions
  • Details:
    SureVector E. coli N-terminal tag Kit E. coli starter kit containing 3 separate N-terminal tags and a T7 promoter