Multiple Affinity Removal Column Human 14, Agilent Technologies
Multiple Affinity Removal Column, Human-14, 4.60 mm, 100 mm
Catalog # CA76193-636
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
: 5188-6558
Multiple Affinity Removal Column Human 14, Agilent Technologies
Catalog # CA76193-636
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Specifications
- Ø int.:4.60 mm
- Désignation:Multiple Affinity Removal Column, Human-14
- Longueur:100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



