Ward's® Sterile Cell Strainers
Product Details & Documents
Ward's® Sterile Cell Strainers and Cell-Straining Kits provide all the necessary parts for filtration of cell suspensions.
- Available as Kits (strainer, 50 ml tube, and cap) or as strainers alone
- Easy to open under the hood, reducing contamination risk
Cell Straining Kits: Eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean users are no longer forced to expose entire bags of 50 ml tubes and corresponding caps in order to perform one filtration. Instead they can open a single packet directly under the hood.
Cell Strainers: If you don't need the accompanying tube that comes with the Cell Straining Kit, you can purchase 40 μm, 70 μm, or 100 μm cell strainers by themselves.
Spécifications
- Taille de pore:100 μm
- Color:Yellow
- Désignation:Large-Mesh
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



