Microscope slides, Kemtech America
76204-208
Kemtech America slides are in compliance with iso 8037/1 international standard in dimension. each slide is pre cleaned and thouroughly examined to ensure premium quality
- Super grade and super white glass available
- 45 degree clipped and 90 degree corners available
- color frosted
- Iso 8037/1 compiant
: ISO Certified: GB/T 19001-2008/ISO 9001:2008
- Cat. No.:PRTWA590WHRL
- Ambidestro/mano specifico:ambidestro
- Materiale:Cotone/viscosa
- Modello:A590
- Materiale rivestimento interno:Cotone
- Tipo polsino:Lungo