Microscope slides, Kemtech America
  76204-208
Microscope slides, Kemtech America
  76204-208
 :  
Microscope slides, Kemtech America
Microscope slides, Kemtech America
Kemtech America slides are in compliance with iso 8037/1 international standard in dimension. each slide is pre cleaned and thouroughly examined to ensure premium quality
  • Super grade and super white glass available
  • 45 degree clipped and 90 degree corners available
  • color frosted
  • Iso 8037/1 compiant
 : ISO Certified: GB/T 19001-2008/ISO 9001:2008
  • Cat. No.:
    PRTWA590WHRL
  • Ambidestro/mano specifico:
    ambidestro
  • Materiale:
    Cotone/viscosa
  • Modello:
    A590
  • Materiale rivestimento interno:
    Cotone
  • Tipo polsino:
    Lungo