StepMate Repeater Pipette
Adapter for 25 mL or 50 mL Syringes
Catalog # 470236-092
Supplier: DLAB SCIENTIFIC INC.
Supplier #: 17400083
Jump to:
Product Details & Documents
Spend less time dispensing student’s reagents with this positive displacement repeating pipettor.
- Dispense up to 48 times without refilling
- Light and ergonomic
- Available volume range from 0.5 to 50 ml
- Excellent for high viscosity liquids
With five dial settings and a full range of syringes available, the repeater pipette allows users to dispense multiple aliquots ranging in volume from 10 to 5000 µl.
Specifications
- Désignation:Adapter for 25 mL or 50 mL Syringes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



