Stirring Rods
Stirring Rod, Glass, 5 mm x 250 mm (10")
Catalog # 470235-782
Supplier:   GSC International
 :  1900-10-DZ
Stirring Rods
Catalog # 470235-782
Supplier:  GSC International
Stirring Rods
Stirring Rods
Stirring Rod, Glass, 5 mm x 250 mm (10")
Price
$6.00
Log in pour la disponibilité
Each Pack of 12
Product Details & Documents
Annealed flint glass rods with fire polished ends.
  • Both ends fire polished and rounded
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.
Spécifications
  • Désignation:
    Stirring Rod, Glass, 5 mm x 250 mm (10")