Eudiometer
Eudiometer, 100 ml
  470235-712
 :  503-2-10
Eudiometer
  470235-712
Eudiometer
Eudiometer
Eudiometer, 100 ml
$77.25
Each Case of 10
Demonstrate the combining volume ratios of gases.
  • Two sizes available
  • Platinum electrodes included
Long, sturdy glass tubes have platinum electrodes sealed at the closed end with a 2 mm arc gap. The tubes are marked with a permanent brown scale in 0.1 or 0.2 mL divisions for 50 and 100 mL respectively. An induction coil is recommended for use with eudiometers.
  • Désignation:
    Eudiometer, 100 ml