Quality Corks, XX
XX Quality Corks, Size 6
Catalog # 470235-706
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CORK XX QUALITY SZ 6 19MM X 14.6MM PK100 Catalog #470310-812
Product 470310-812 is a replacement for 470235-706 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Size Top Diameter Bottom Diameter
Size 1 11.1 mm 8.3 mm
Size 2 12.7 mm 9.5 mm
Size 3 14.2 mm 10.7 mm
Size 4 15.8 mm 11.9 mm
Size 6 19.0 mm 14.6 mm
Size 20 41.2 mm 34.0 mm
- Size 2 - sold in a case; 10 packs of 100
- Size 3 - sold in a case; 10 packs of 100
- Size 4 - sold in a case; 10 packs of 100
- Size 20 - sold in a pack of 10
- The natural cork is safe for use with food products
Get the perfect fit with our lab-grade cork stoppers. These stoppers ensure a secure, leak-proof seal every time. Crafted from natural cork, these stoppers are safe for use with food products, making them a reliable choice for all your storage needs.
Specifications
- Désignation:XX Quality Corks, Size 6